Tetradecanoic acid, 98%
Tetradecanoic acid, 98%
Tetradecanoic acid, 98%
Thermo Scientific Chemicals

Tetradecanoic acid, 98%

CAS: 544-63-8 | C14H28O2 | 228.38 g/mol
数量:
250 g
500 g
2500 g
製品番号(カタログ番号) A12067.0E
または、製品番号A12067-0E
価格(JPY)
-
数量:
2500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS544-63-8
Melting Point (clear melt)52.0-58.0?C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Appearance (Color)White
Assay (Silylated GC)≥97.5%
さらに表示
It is employed as a flow Agent and emulsifier. Used in artificial orris fragrances. Employed as anionic/nonionic Surfactants in soaps and detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a flow Agent and emulsifier. Used in artificial orris fragrances. Employed as anionic/nonionic Surfactants in soaps and detergents.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
RUO – Research Use Only

General References:

  1. Erika M Hernández.; Tze-Lee Phang.; Xinyun Wen.; Elias I Franses. Adsorption and direct probing of fibrinogen and sodium myristate at the air/water interface.J Colloid Interface Sci. 2002,250(2), 271-280.
  2. Wen X.; Franses EI. Effect of Protonation on the Solution and Phase Behavior of Aqueous Sodium Myristate.J Colloid Interface Sci. 2000,23142-51.