Tyramine hydrochloride, 98%
Tyramine hydrochloride, 98%
Tyramine hydrochloride, 98%
Thermo Scientific Chemicals

Tyramine hydrochloride, 98%

CAS: 60-19-5 | C8H12ClNO | 173.64 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A12220.06
または、製品番号A12220-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS60-19-5
IUPAC Namehydrogen 4-(2-aminoethyl)phenol chloride
Molecular FormulaC8H12ClNO
InChI KeyRNISDHSYKZAWOK-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCCC1=CC=C(O)C=C1
さらに表示
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Melting Point (clear melt)271.0-277.0?C
Reagent for fluorescence enzyme immunoassay

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only