2,3-Dichloropyridine, 99%
2,3-Dichloropyridine, 99%
2,3-Dichloropyridine, 99%
Thermo Scientific Chemicals

2,3-Dichloropyridine, 99%

CAS: 2402-77-9 | C5H3Cl2N | 147.99 g/mol
製品番号(カタログ番号) A12241.14
または、製品番号A12241-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2402-77-9
IUPAC Name2,3-dichloropyridine
Molecular FormulaC5H3Cl2N
InChI KeyMAKFMOSBBNKPMS-UHFFFAOYSA-N
SMILESClC1=CC=CN=C1Cl
さらに表示
Appearance (Color)White to pale cream to pale brown
Melting Point (clear melt)64.0-71.0?C
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder or fused solid or flakes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only