Benzo[b]thiophene-2-carboxylic acid, 98%
Benzo[b]thiophene-2-carboxylic acid, 98%
Benzo[b]thiophene-2-carboxylic acid, 98%
Thermo Scientific Chemicals

Benzo[b]thiophene-2-carboxylic acid, 98%

CAS: 6314-28-9 | C9H6O2S | 178.21 g/mol
製品番号(カタログ番号) A12471.03
または、製品番号A12471-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6314-28-9
Assay (Aqueous acid-base Titration)>97.5 to <102.5%
Appearance (Color)White to cream
FormPowder
Benzo[b]thiophene-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in ethanol, acetone and chloroform. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Arun M. Isloor.; Balakrishna Kalluraya.; K. Sridhar Pai. Synthesis, characterization and biological activities of some new benzo[b]thiophene derivatives. European Journal of Medicinal Chemistry. 2010, 45 (2), 825-830.
  2. Bernd Peschke.; Sonja Bak.; Rolf Hohlweg.; Rita Nielsen.; Dorthe Viuff.; Karin Rimvall. Benzo[b]thiophene-2-carboxamides and benzo[b]furan-2-carboxamides are potent antagonists of the human H3-receptor. Bioorganic & Medicinal Chemistry Letters. 2006, 16 (12), 3162-3165.