Glycolic acid, 98%
Glycolic acid, 98%
Glycolic acid, 98%
Thermo Scientific Chemicals

Glycolic acid, 98%

CAS: 79-14-1 | C2H4O3 | 76.051 g/mol
製品番号(カタログ番号) A12511.22
または、製品番号A12511-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS79-14-1
IUPAC Name2-hydroxyacetic acid
Molecular FormulaC2H4O3
InChI KeyAEMRFAOFKBGASW-UHFFFAOYSA-N
SMILESOCC(O)=O
さらに表示
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤2%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Glycolic acid is used for acne and anti-aging treatment. It is also employed in the textile industry as a dyeing and tanning agent. It is used in emulsion polymers, solvents and additives for ink and paints. It is also useful as an intermediate for organic synthesis such as redox reactions, esterification and long chain polymerization. It acts as a monomer in the preparation of polyglycolic acid and other biocompatible copolymers. Further, it is used as a preservative and flavoring agent in organic synthesis. It is also used in adhesives and plastics.

Solubility
Soluble in water, methanol, diethyl ether, acetone, acetic acid and ethyl acetate.

Notes
Hygroscopic. Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

General References:

  1. Datta, D.; Kumar, S. Modeling Using Response Surface Methodology And Optimization Using Differential Evolution Of Reactive Extraction Of Glycolic Acid. Chem. Eng. Commun. 2015, 202 (1), 59-69.
  2. Steward, K. M.; Corbett, M. T.; Goodman, C. G.; Johnson, J. S. Correction to Asymmetric Synthesis of Diverse Glycolic Acid Scaffolds via Dynamic Kinetic Resolution of α-Keto Esters. J. Am. Chem. Soc. 2015, 137 (11), 3991-3991.