Thermo Scientific Chemicals

2,4-Dimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A12549.14
25 g, Each
Thermo Scientific Chemicals

2,4-Dimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A12549.14
25 g, Each
数量
製品番号(カタログ番号): A12549.14
または、製品番号A12549-14

化学物質識別子

CAS
613-45-6
IUPAC Name
2,4-dimethoxybenzaldehyde
Molecular Formula
C9H10O3
InChI Key
LWRSYTXEQUUTKW-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=O)C(OC)=C1
Assay (GC)
≥97.5%
Melting Point (clear melt)
68-72?C
Appearance (Color)
White to pale yellow or cream to pale brown
Form
Crystals or powder or crystalline powder

概要

2,4-Dimethoxybenzaldehyde is used as a reagent to specifically quantify phlorotannins. It is also used as a component of hair dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dimethoxybenzaldehyde is used as a reagent to specifically quantify phlorotannins. It is also used as a component of hair dyes.

Solubility
Insoluble in water. Soluble in chlorofrom.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text