Naphthalene-2-sulfonic acid sodium salt, 98%, may cont. up to 10% residual inorganic salts and water
Naphthalene-2-sulfonic acid sodium salt, 98%, may cont. up to 10% residual inorganic salts and water
Naphthalene-2-sulfonic acid sodium salt, 98%, may cont. up to 10% residual inorganic salts and water
Thermo Scientific Chemicals

Naphthalene-2-sulfonic acid sodium salt, 98%, may cont. up to 10% residual inorganic salts and water

CAS: 532-02-5 | C10H7NaO3S | 230.21 g/mol
製品番号(カタログ番号) A13066.30
または、製品番号A13066-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS532-02-5
IUPAC Namesodium naphthalene-2-sulfonate
Molecular FormulaC10H7NaO3S
InChI KeyYWPOLRBWRRKLMW-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C1=CC=C2C=CC=CC2=C1
さらに表示
Appearance (Color)White to pale brown
FormPowder
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
Impurity contentSodium chloride, NaCl <7.0%
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only