2-(1-Piperidinyl)aniline, 98%
Thermo Scientific Chemicals

2-(1-Piperidinyl)aniline, 98%

製品番号(カタログ番号)数量
A13073.03
または、製品番号A13073-03
1 g
製品番号(カタログ番号) A13073.03
または、製品番号A13073-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS39643-31-7
IUPAC Name2-(piperidin-1-yl)aniline
Molecular FormulaC11H16N2
InChI KeyOYECAJPUPWFCSL-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1N1CCCCC1
さらに表示
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Appearance (Color)Brown to dark brown
Melting Point (clear melt)43.0-49.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.