Thermo Scientific Chemicals

3-Methylthiophene-2-carbonitrile, 96%, Thermo Scientific Chemicals

製品番号(カタログ番号): A13305.03
1 g, Each
Thermo Scientific Chemicals

3-Methylthiophene-2-carbonitrile, 96%, Thermo Scientific Chemicals

製品番号(カタログ番号): A13305.03
1 g, Each
数量
製品番号(カタログ番号): A13305.03
または、製品番号A13305-03

化学物質識別子

CAS
55406-13-8
IUPAC Name
3-methylthiophene-2-carbonitrile
Molecular Formula
C6H5NS
InChI Key
ALZHYEITUZEZMT-UHFFFAOYSA-N
SMILES
CC1=C(SC=C1)C#N
Refractive Index
1.5505-1.5555 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥95.0%
Form
Liquid

概要

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text