6-Methyl-5-hepten-2-one, 98%
6-Methyl-5-hepten-2-one, 98%
6-Methyl-5-hepten-2-one, 98%
Thermo Scientific Chemicals

6-Methyl-5-hepten-2-one, 98%

CAS: 110-93-0 | C8H14O | 126.20 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A13335.14
または、製品番号A13335-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS110-93-0
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4380-1.4420 @ 20?C
6-Methyl-hepten-2-one is used in the synthesis of thyrsiferyl 23-Acetate which acts as an anti-leukemic inducer of apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-hepten-2-one is used in the synthesis of thyrsiferyl 23-Acetate which acts as an anti-leukemic inducer of apoptosis.

Solubility
Soluble in methanol and chloroform.

Notes
Store in cool, dry place in tightly closed container.
RUO – Research Use Only

General References:

  1. Charles J Weschler et. al. Ozone-initiated chemistry in an occupied simulated aircraft cabin. Environmental Science & Technology,. 2007, 41 (17), 6177-6184.
  2. W A Wolken et. al. Amino acid-catalyzed conversion of citral: cis-trans isomerization and its conversion into 6-methyl-5-hepten-2-one and acetaldehyde. Journal of Agricultural and Food Chemistry,. 2000, 48 (11), 5401-5405.