3-Methylthiophene, 98+%
3-Methylthiophene, 98+%
3-Methylthiophene, 98+%
Thermo Scientific Chemicals

3-Methylthiophene, 98+%

CAS: 616-44-4 | C5H6S | 98.163 g/mol
製品番号(カタログ番号) A13394.14
または、製品番号A13394-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS616-44-4
IUPAC Name3-methylthiophene
Molecular FormulaC5H6S
InChI KeyQENGPZGAWFQWCZ-UHFFFAOYSA-N
SMILESCC1=CSC=C1
さらに表示
Refractive Index1.5180-1.5220 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only