Quinaldine, 96%
Quinaldine, 96%
Quinaldine, 96%
Thermo Scientific Chemicals

Quinaldine, 96%

CAS: 91-63-4 | C10H9N | 143.19 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A13412.22
または、製品番号A13412-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS91-63-4
IUPAC Name2-methylquinoline
Molecular FormulaC10H9N
InChI KeySMUQFGGVLNAIOZ-UHFFFAOYSA-N
SMILESCC1=CC=C2C=CC=CC2=N1
さらに表示
Appearance (Color)Clear colorless to yellow to brown
FormLiquid
Refractive Index1.6080-1.6145 @ 20?C
Assay (GC)≥95.0%
It is an anaesthetic used in fish transportation. Used in preparation of oil-soluble dyes. Also used in anti-malaria drugs, in manufacturing dyes, food colorants , pharmaceuticals and pH indicators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an anaesthetic used in fish transportation. Used in preparation of oil-soluble dyes. Also used in anti-malaria drugs, in manufacturing dyes, food colorants , pharmaceuticals and pH indicators.

Solubility
Soluble in water (2.5g/L).

Notes
Air and Light Sensitive. Store in dark and away from air. Store in cool, dry place in tightly closed container. Store away from acid chlorides and acid anhydrides. It may discolor to brown upon storage.
RUO – Research Use Only

General References:

  1. R. N. Gibson. The Use of the Anaesthetic Quinaldine in Fish Ecology. Journal of Animal Ecology. 1967, 36 (2), 295-301,.
  2. D Ferri; and T Bürgi. An in situ attenuated total reflection infrared study of a chiral catalytic solid-liquid interface: cinchonidine adsorption on pt. Journal of the American Chemical Society, 2001, 123 (48), 12074-12084.