9-(Chloromethyl)anthracene, 98+%
9-(Chloromethyl)anthracene, 98+%
9-(Chloromethyl)anthracene, 98+%
Thermo Scientific Chemicals

9-(Chloromethyl)anthracene, 98+%

CAS: 24463-19-2 | C15H11Cl | 226.703 g/mol
製品番号(カタログ番号) A13579.03
または、製品番号A13579-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS24463-19-2
IUPAC Name9-(chloromethyl)anthracene
Molecular FormulaC15H11Cl
InChI KeyPCVRSXXPGXRVEZ-UHFFFAOYSA-N
SMILESClCC1=C2C=CC=CC2=CC2=CC=CC=C12
さらに表示
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Identification (FTIR)Conforms
9-(Chloromethyl)anthracene is used as a blocking group reagent used for carboxylic acids, phenols, mercaptans and thiophenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-(Chloromethyl)anthracene is used as a blocking group reagent used for carboxylic acids, phenols, mercaptans and thiophenols.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Korte, W.D. 9(chloromethyl)anthracene: a useful derivatizing reagent for enhanced ultraviolet and fluorescence detection of carboxylic acids with liquid chromatography. Journal of Chromatography A. 1982, 243, (1), 153-157.
  2. Reagent for the protection of carboxylic acids, alcohols, phenols and thiols as their 9-anthrylmethyl derivatives, which are stable to mild acid or base, but specifically cleaved by NaSMe: J. Am. Chem. Soc., 96, 590 (1974). For use in N-protection, see: Aust. J. Chem., 18, 1699 (1965).