2-Bromobenzoic acid, 98%
2-Bromobenzoic acid, 98%
2-Bromobenzoic acid, 98%
Thermo Scientific Chemicals

2-Bromobenzoic acid, 98%

CAS: 88-65-3 | C7H5BrO2 | 201.019 g/mol
数量:
50 g
250 g
1000 g
製品番号(カタログ番号) A13608.18
または、製品番号A13608-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS88-65-3
IUPAC Name2-bromobenzoic acid
Molecular FormulaC7H5BrO2
InChI KeyXRXMNWGCKISMOH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1Br
さらに表示
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)146.0-152.0?C
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only