Ammonium molybdate (para) tetrahydrate, 99%
Ammonium molybdate (para) tetrahydrate, 99%
Ammonium molybdate (para) tetrahydrate, 99%
Thermo Scientific Chemicals

Ammonium molybdate (para) tetrahydrate, 99%

CAS: 12054-85-2 | (NH4)6Mo7O24·4H2O
数量:
50 g
250 g
1000 g
製品番号(カタログ番号) A13766.18
または、製品番号A13766-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12054-85-2
IUPAC Nameammonium hydrate molybdenum oxidandiide
Molecular FormulaH6MoNO2
InChI KeyXCOMAVCHADVANQ-UHFFFAOYSA-O
SMILES[NH4+].O.[O--].[Mo]
さらに表示
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Solution TestWater, 5%w/v, 30mins: Clear
Assay (Titration)≥98.5 to ≤101.5%
Ammonium molybdate tetrahydrate is widely utilized in cryo-negative staining which helps to visualize particles while preserving the sample. It is used as a catalyst to desulfurize thiophene and its derivatives. It serves as an analytical reagent to measure the amount of phosphates, silicates, arsenates and lead in an aqueous solution. It is also used in electroplating, in fertilizers for crops and in the production of molybdenum metal and ceramics. It is used as an analgesic in medical field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium molybdate tetrahydrate is widely utilized in cryo-negative staining which helps to visualize particles while preserving the sample. It is used as a catalyst to desulfurize thiophene and its derivatives. It serves as an analytical reagent to measure the amount of phosphates, silicates, arsenates and lead in an aqueous solution. It is also used in electroplating, in fertilizers for crops and in the production of molybdenum metal and ceramics. It is used as an analgesic in medical field.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. The combination with hydrogen peroxide is a system for the ambient temperature oxidative cleavage of epoxides to form ɑ-hydroxy ketones: Chem. Lett., 844 (2000).
  2. Lince, J. R.; Pluntze, A. M.; Jackson, A.; Radhakrishnan, G.; Adams, P. M. Tribochemistry of MoS3Nanoparticle Coatings. Tribol Lett. 2014, 53 (3), 543-554.
  3. McIntosha, S.; Bosscheb, M. V. Towards Understanding the Hydrocarbon Oxidation Activity of Oxides for Direct Hydrocarbon SOFC Anodes. ECS Trans. 2011, 35 (1), 2781-2788.