(1-Hexyl)triphenylphosphonium bromide, 98%
(1-Hexyl)triphenylphosphonium bromide, 98%
(1-Hexyl)triphenylphosphonium bromide, 98%
Thermo Scientific Chemicals

(1-Hexyl)triphenylphosphonium bromide, 98%

CAS: 4762-26-9 | C24H28BrP | 427.37 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A13826.14
または、製品番号A13826-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4762-26-9
IUPAC Namehexyltriphenylphosphanium bromide
Molecular FormulaC24H28BrP
InChI KeyPWDFZWZPWFYFTC-UHFFFAOYSA-M
SMILES[Br-].CCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
FormPowder or lumps
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
Melting Point (clear melt)198.0-205.0?C
Appearance (Color)White to pale cream to pale yellow
Water Content (Karl Fischer Titration)≤1%
(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Store under inert gas. Protect from light. Store away from oxidizing materials.
RUO – Research Use Only