Phenyltrimethoxysilane, 97%
Phenyltrimethoxysilane, 97%
Phenyltrimethoxysilane, 97%
Thermo Scientific Chemicals

Phenyltrimethoxysilane, 97%

CAS: 2996-92-1 | C9H14O3Si | 198.29 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A13827.36
または、製品番号A13827-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2996-92-1
IUPAC Nametrimethoxy(phenyl)silane
Molecular FormulaC9H14O3Si
InChI KeyZNOCGWVLWPVKAO-UHFFFAOYSA-N
SMILESCO[Si](OC)(OC)C1=CC=CC=C1
さらに表示
Identification (FTIR)Conforms
Refractive Index1.4700-1.4750 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Phenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Desiccate at room temperature. Store under inert gas. Incompatible with Strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Touria Jermouni; Monique Smaihi and Nadine Hovnanian. Hydrolysis and initial polycondensation of phenyltrimethoxysilane and diphenyldimethoxysilane.J. Mater. Chem.1995, 5 1203-1208.
  2. Yung-Chih Chen; Hsueh-Chen Lin; Yu-Der Lee. The Effects of Phenyltrimethoxysilane Coupling Agents on the Properties of PTFE/Silica Composites.Journal of Polymer Research.2004, 11 1-7
  3. Aryltrimethoxysilanes undergo Pd-catalyzed cross-coupling reactions with e.g. aryl iodides in the presence of TBAF, via a postulated hypervalent siloxane intermediate: J. Org. Chem., 64, 1684 (1999):