Thermo Scientific Chemicals

6-Aza-2-thiothymine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A14167.22
100 g, Each
Thermo Scientific Chemicals

6-Aza-2-thiothymine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A14167.22
100 g, Each
数量
製品番号(カタログ番号): A14167.22
または、製品番号A14167-22

化学物質識別子

CAS
615-76-9
IUPAC Name
6-methyl-3-sulfanylidene-2,3,4,5-tetrahydro-1,2,4-triazin-5-one
Molecular Formula
C4H5N3OS
InChI Key
NKOPQOSBROLOFP-UHFFFAOYSA-N
SMILES
CC1=NNC(=S)NC1=O
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥97.5 to ≤102.5% (titration)

概要

6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.

Solubility
Soluble in water (partly), and formic acid (5 %).

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text