2-Chloro-5-nitrobenzyl alcohol, 98%
2-Chloro-5-nitrobenzyl alcohol, 98%
2-Chloro-5-nitrobenzyl alcohol, 98%
Thermo Scientific Chemicals

2-Chloro-5-nitrobenzyl alcohol, 98%

CAS: 80866-80-4 | C7H6ClNO3 | 187.579 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A14205.22
または、製品番号A14205-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS80866-80-4
IUPAC Name(2-chloro-5-nitrophenyl)methanol
Molecular FormulaC7H6ClNO3
InChI KeyNNLQYDLTFRXAKD-UHFFFAOYSA-N
SMILESOCC1=CC(=CC=C1Cl)[N+]([O-])=O
さらに表示
Appearance (Color)Cream to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)71.0-78.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only