Tetramethyldisilazane, 97%
Tetramethyldisilazane, 97%
Tetramethyldisilazane, 97%
Thermo Scientific Chemicals

Tetramethyldisilazane, 97%

CAS: 15933-59-2 | C4H13NSi2 | 131.325 g/mol
製品番号(カタログ番号) A14304.18
または、製品番号A14304-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS15933-59-2
Molecular FormulaC4H13NSi2
InChI KeyGJWAPAVRQYYSTK-UHFFFAOYSA-N
SMILESC[Si](C)N[Si](C)C
Molecular Weight (g/mol)131.33
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay from Supplier's CofA≥96.0% (GC)
FormLiquid
Tetramethyldisilazane is used as a gas chromatographic derivatizing reagent. Further, it reacts with phenol to prepare dimethylphenoxysilane. In addition, it is used in electronic, polymer and pharmaceutical industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyldisilazane is used as a gas chromatographic derivatizing reagent. Further, it reacts with phenol to prepare dimethylphenoxysilane. In addition, it is used in electronic, polymer and pharmaceutical industries.

Solubility
Miscible with common organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Reagent for the preparation (see Appendix 4) of dimethylsilyl (DMS) derivatives, compare Hexamethyl disilazane, A15139. With allylic and homoallylic alcohols, the products can undergo Pt- or Rh-catalyzed intramolecular hydrosilylation, providing a regioselective route to 1,3-diols: J. Am. Chem. Soc., 108, 6090 (1986). For an example of this reaction, see Chlorodimethyl silane, A13113.
  2. The potassium derivative, formed by reaction with KH in THF, reacts with alkyl halides to give, after hydrolysis, high yields of primary amines, thus providing a convenient alternative to the classical Gabriel method. Other silazanes, including hexamethyldisilazane, give lower yields: Synthesis, 150 (1995):
  3. Sanli, D.; Erkey, C. Silylation from supercritical carbon dioxide: a powerful technique for modification of surfaces. J. Mater. Sci. 2015, 50 (22), 7159-7181.
  4. Fainer, N. I.; Kosyakov, V. I. Phase composition of thin silicon carbonitride films obtained by plazma endanced chemical vapor deposition using organosilicon compounds. J. Struct. Chem. 2015, 56 (1), 163-174.