Thermo Scientific Chemicals

4-Benzylpyridine, 97+%, Thermo Scientific Chemicals

製品番号(カタログ番号): A14326.36
500 g, Each
Thermo Scientific Chemicals

4-Benzylpyridine, 97+%, Thermo Scientific Chemicals

製品番号(カタログ番号): A14326.36
500 g, Each
数量
製品番号(カタログ番号): A14326.36
または、製品番号A14326-36

化学物質識別子

CAS
2116-65-6
IUPAC Name
4-benzylpyridine
Molecular Formula
C12H11N
InChI Key
DBOLXXRVIFGDTI-UHFFFAOYSA-N
SMILES
C(C1=CC=CC=C1)C1=CC=NC=C1
Appearance (Color)
Clear, colorless to pale yellow or pale brown
Form
Liquid
Assay (GC)
≥97.0%
Identification (FTIR)
Conforms
Refractive Index
1.5785-1.5835 @ 20?C

概要

4-Benzylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in ethanol, ethyl ether. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong acids.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text