1-Bromoundecane, 98%
1-Bromoundecane, 98%
1-Bromoundecane, 98%
Thermo Scientific Chemicals

1-Bromoundecane, 98%

CAS: 693-67-4 | C11H23Br | 235.209 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A14333.22
または、製品番号A14333-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS693-67-4
IUPAC Name1-bromoundecane
Molecular FormulaC11H23Br
InChI KeyIKPSIIAXIDAQLG-UHFFFAOYSA-N
SMILESCCCCCCCCCCCBr
さらに表示
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Refractive Index1.4550-1.4590 @ 20?C
FormLiquid
Assay (GC)≥97.5%
1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromoundecane is used in the preparation of Grignard reagent by reacting with Mg in THF (tetrahydrofuran). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Kumaraswamy G.; Markondaiah B. Enantioselective total synthesis of (-)-tetrahydrolipstatin using Oppolzer’s sultam directed aldol reaction. Tetrahedron Lett. 2008, 49 (2), 327-330.
  2. Jean-François Peyrata.; Bruno Figadèrea.; André Cavé. Halide Exchange: Preparation of Alkyl Chlorides. Synthetic Communications: An International Journal for Rapid Communication of Synthetic Organic Chemistry. 1996, 26 (24), 4563-4567.