2',6'-Dihydroxyacetophenone, 98+%
2',6'-Dihydroxyacetophenone, 98+%
2',6'-Dihydroxyacetophenone, 98+%
Thermo Scientific Chemicals

2',6'-Dihydroxyacetophenone, 98+%

CAS: 699-83-2 | C8H8O3 | 152.149 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A14627.14
または、製品番号A14627-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS699-83-2
IUPAC Name1-(2,6-dihydroxyphenyl)ethan-1-one
Molecular FormulaC8H8O3
InChI KeyYPTJKHVBDCRKNF-UHFFFAOYSA-N
SMILESCC(=O)C1=C(O)C=CC=C1O
さらに表示
Assay (GC)≥98.0%
Appearance (Color)Pale cream to cream to yellow to pale brown
Melting Point (clear melt)154.0-161.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only