Allyltrimethylsilane, 98+%
Allyltrimethylsilane, 98+%
Allyltrimethylsilane, 98+%
Thermo Scientific Chemicals

Allyltrimethylsilane, 98+%

CAS: 762-72-1 | C6H14Si | 114.263 g/mol
製品番号(カタログ番号) A14662.09
または、製品番号A14662-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS762-72-1
IUPAC Nametrimethyl(prop-2-en-1-yl)silane
Molecular FormulaC6H14Si
InChI KeyHYWCXWRMUZYRPH-UHFFFAOYSA-N
SMILESC[Si](C)(C)CC=C
さらに表示
Appearance (Color)Clear colorless
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4060-1.4100 @ 20?C
Assay (GC)≥98.0%
Allyltrimethylsilane is involved as a reactant in Hosomi Sakurai reaction for allylation in the presence of Lewis acid. For example, it reacts with cyclohexanone to get 1-allylcyclohexanol. It acts as a nucleophile and is involved in Carbon-Ferrier rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyltrimethylsilane is involved as a reactant in Hosomi Sakurai reaction for allylation in the presence of Lewis acid. For example, it reacts with cyclohexanone to get 1-allylcyclohexanol. It acts as a nucleophile and is involved in Carbon-Ferrier rearrangement.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents and acids.
RUO – Research Use Only

General References:

  1. Reactions with electrophiles illustrate the ß-effect (see Appendix 4); e.g. acid chlorides with a Lewis acid give allyl ketones: J. Organomet. Chem., 85, 149 (1975):
  2. Similarly, carbonyl compounds are converted to homoallylic alcohols: J. Organomet. Chem., 69, C15 (1974). In the TiCl4 promoted reaction with enones, behaves as an allyl anion equivalent, giving the product of conjugate addition (Hosomi-Sakurai reaction): J. Am. Chem. Soc., 99, 1673 (1977); 105, 2354 (1983); Org. Synth. Coll., 7, 443 (1990):
  3. More recent studies have shown that TMS cyclopentanes are also formed in these reactions via a competing [3+2] cycloaddition mechanism: Tetrahedron, 49, 9955 (1993). Chemoselective addition to aldehydes can be accomplished in the presence of a ketone, using Scandium(III) trifluoromethanesulfonate hydrate, 40566 as catalyst: Synthesis, 1822 (1998).
  4. In the presence of F-, functions as an allyl anion equivalent, affording homoallylic alcohols from carbonyl compounds: Tetrahedron Lett., 3043 (1978), and adding to a variety of Michael acceptors: Tetrahedron Lett., 25, 3213 (1984); J. Org. Chem., 51, 1745 (1986). For a review of reactivity in the presence of Lewis acids and F-, see: J. Prakt. Chem./ Chem. Ztg., 336, 375 (1994).
  5. Has also been used, in the presence of a catalyst such as p-TsOH, I2, Br2 or TfOH, as a silylating agent for alcohols, phenols and carboxylic acids: Tetrahedron Lett., 21, 835 (1980); J. Org. Chem., 46, 5212 (1981). The advantage over more conventional silylation methods is that the only by-product is the neutral gas propene.