Adipic dihydrazide, 97%
Adipic dihydrazide, 97%
Adipic dihydrazide, 97%
Thermo Scientific Chemicals

Adipic dihydrazide, 97%

CAS: 1071-93-8 | C6H14N4O2 | 174.20 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A15119.14
または、製品番号A15119-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1071-93-8
IUPAC Namehexanedihydrazide
Molecular FormulaC6H14N4O2
InChI KeyIBVAQQYNSHJXBV-UHFFFAOYSA-N
SMILESNNC(=O)CCCCC(=O)NN
さらに表示
Appearance (Color)White to pale cream or pale yellow
FormPowder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥96.0 to <104.0%
Melting Point (clear melt)178.0-184.0?C
Adipic dihydrazide is used as a formaldehyde scavenger and reacts with formaldehyde, thereby preventing the volatilizing of formaldehyde in the air. It is also employed as a paint additive and coating additive. It is also used as an intermediate. Further, it is used for cross-linking water-based emulsions and as a hardener for certain epoxy resins, which finds application in powder coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adipic dihydrazide is used as a formaldehyde scavenger and reacts with formaldehyde, thereby preventing the volatilizing of formaldehyde in the air. It is also employed as a paint additive and coating additive. It is also used as an intermediate. Further, it is used for cross-linking water-based emulsions and as a hardener for certain epoxy resins, which finds application in powder coating.

Solubility
Soluble in water, alcohol and acetone.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Xie, D.; Liao, K.; Wang, L.; Zhang, X. Behavior of adipic dihydrazide and silica in the preparation of acrylate redispersible polymer powders. Colloid. Polym. Sci. 2015, 293 (7), 1937-1944.
  2. Pi, P.; Wang, W.; Wen, X.; Xu, S.; Cheng, J. Synthesis and characterization of low-temperature self-crosslinkable acrylic emulsion for PE film ink. Prog. Org. Coat. 2015, 81, 66-71.