Thermo Scientific Chemicals

Trimethyloxonium tetrafluoroborate, 96%, Thermo Scientific Chemicals

製品番号(カタログ番号): A15175.18
50 g, Each
Thermo Scientific Chemicals

Trimethyloxonium tetrafluoroborate, 96%, Thermo Scientific Chemicals

製品番号(カタログ番号): A15175.18
50 g, Each
数量
製品番号(カタログ番号): A15175.18
または、製品番号A15175-18

化学物質識別子

CAS
420-37-1
IUPAC Name
tetrafluoroboranuide; trimethyloxidanium
Molecular Formula
C3H9BF4O
InChI Key
CZVZBKHWOFJNCR-UHFFFAOYSA-N
SMILES
C[O+](C)C.F[B-](F)(F)F
Appearance (Color)
White to pale cream
Form
Powder or lumps
Assay (Aqueous acid-base Titration)
>95.0%
Solution Test
Clear (2.5% w/v in water)

概要

Trimethyloxonium tetrafluoroborate act as a avmethylating agent and activates C-X multiple bonds. It is involved in the esterification of polyfunctional carboxylic acids. It acts as a catalyst in the polymerization of cyclic sulfides and ethers. Further, it is used in Beckmann rearrangement of oximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethyloxonium tetrafluoroborate act as a avmethylating agent and activates C-X multiple bonds. It is involved in the esterification of polyfunctional carboxylic acids. It acts as a catalyst in the polymerization of cyclic sulfides and ethers. Further, it is used in Beckmann rearrangement of oximes.

Solubility
Soluble in nitrobenzene, nitromethane, chloroform, hot acetone, liquid sulfur dioxide. Slightly soluble in dichloromethane. Insoluble in common organic solvents.

Notes
Hygroscopic. Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text