9-Anthracenemethanol, 97%
9-Anthracenemethanol, 97%
9-Anthracenemethanol, 97%
Thermo Scientific Chemicals

9-Anthracenemethanol, 97%

CAS: 1468-95-7 | C15H12O | 208.26 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) A15188.09
または、製品番号A15188-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1468-95-7
IUPAC Name(anthracen-9-yl)methanol
Molecular FormulaC15H12O
InChI KeyJCJNNHDZTLRSGN-UHFFFAOYSA-N
SMILESOCC1=C2C=CC=CC2=CC2=CC=CC=C12
さらに表示
FormCrystals or powder or crystalline powder
Appearance (Color)Yellow to green
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)156.0-167.0?C
An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An anthracene derivative used in Diels-Alder reactions. Also used in the preparation of 9-anthracenylmethyl-1-piperazinecarboxylate.

Solubility
Soluble in hot methanol very faint turbidity.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Roh Y-M, et al. Development of a new approach for total isocyanate determination using the reagent 9-anthracenylmethyl-1-piperazinecarboxylate. Analyst, 2000, 125(9), 1691-96.
  2. Chen-Yu Li; Chen-Yen Tsai; Chia-Her Lin; Bao-Tsan Ko. Synthesis, structural characterization and reactivity of aluminum complexes supported by benzotriazole phenoxide ligands: air-stable alumoxane as an efficient catalyst for ring-opening polymerization of L-lactide. Dalton Transactions (Print Edition), 2011, 40(9), 1880-1887.
  3. For use of the 9-anthrylmethyl group for carboxyl protection, see 9-(Chloromethyl) anthracene, A13579.