2-Acetyl-1-methylpyrrole, 98%
2-Acetyl-1-methylpyrrole, 98%
2-Acetyl-1-methylpyrrole, 98%
Thermo Scientific Chemicals

2-Acetyl-1-methylpyrrole, 98%

CAS: 932-16-1 | C7H9NO | 123.16 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A15353.06
または、製品番号A15353-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS932-16-1
IUPAC Name1-(1-methyl-1H-pyrrol-2-yl)ethan-1-one
Molecular FormulaC7H9NO
InChI KeyNZFLWVDXYUGFAV-UHFFFAOYSA-N
SMILESCN1C=CC=C1C(C)=O
さらに表示
Appearance (Color)Clear colorless to yellow to orange
Assay (GC)≥97.5%
Refractive Index1.5390-1.5430 @ 20°C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only