Thermo Scientific Chemicals

4-Fluorobenzaldehyde, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A15383.14
25 g, Each
Thermo Scientific Chemicals

4-Fluorobenzaldehyde, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A15383.14
25 g, Each
数量
製品番号(カタログ番号): A15383.14
または、製品番号A15383-14

化学物質識別子

CAS
459-57-4
IUPAC Name
4-fluorobenzaldehyde
Molecular Formula
C7H5FO
InChI Key
UOQXIWFBQSVDPP-UHFFFAOYSA-N
SMILES
FC1=CC=C(C=O)C=C1
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Refractive Index
1.5190-1.5230 @ 20?C
Form
Liquid

概要

4-Fluorobenzaldehyde is used in the preparation of pyrazolopyridine derivative, which finds application as mitogen-activated protein kinase (MAPK) inhibitor. It also serves as a synthetic intermediate in the preparation of pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzaldehyde is used in the preparation of pyrazolopyridine derivative, which finds application as mitogen-activated protein kinase (MAPK) inhibitor. It also serves as a synthetic intermediate in the preparation of pharmaceutical compounds.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text