2-Cyanothioacetamide, 98%
2-Cyanothioacetamide, 98%
2-Cyanothioacetamide, 98%
2-Cyanothioacetamide, 98%
Thermo Scientific Chemicals

2-Cyanothioacetamide, 98%

CAS: 7357-70-2 | C3H4N2S | 100.14 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A15436.14
または、製品番号A15436-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7357-70-2
IUPAC Name2-cyanoethanethioamide
Molecular FormulaC3H4N2S
InChI KeyBHPYMZQTCPRLNR-UHFFFAOYSA-N
SMILESNC(=S)CC#N
さらに表示
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)110-120°C
Appearance (Color)Yellow-orange to brown
FormCrystals or powder or crystalline powder
2-Cyanothioacetamide was used in the synthesis of 3,4-trans-4-aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates and bis[6-(2-aryl)-2-thioxo-1,2-dihydropyridine-3-carbonitrile]. 2-Cyanothioacetamide is used as a building block for 2-pyridothiones. It can be used in agrochemical, pharmaceutical and dyestuff field etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanothioacetamide was used in the synthesis of 3,4-trans-4-aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates and bis[6-(2-aryl)-2-thioxo-1,2-dihydropyridine-3-carbonitrile]. 2-Cyanothioacetamide is used as a building block for 2-pyridothiones. It can be used in agrochemical, pharmaceutical and dyestuff field etc,.

Solubility
Does not mix well with water.

Notes
Light Sensitive. Store in the dark.
RUO – Research Use Only

General References:

  1. Elgemeie G. E. H. ; Ali H . A. ; Eid M. M. Reaction of 2-cyanothioacetamide with α-alkylated β-diketones: synthesis of pyridine-2(1H)-thione, thieno[2,3-b]pyridine and pyrido[2,3-c]pyrazole derivatives. Journal of Chemical Research. Synopses.1993, 7 256-257.
  2. Cyclocondensation with Benzoyl isothiocyanate, L06964 results in the formation of a pyrimidinedithiol: Chem. Ind. (London), 270 (1988):
  3. See also Methyl isothiocyanate, A11757.
  4. For a review on the utility of cyanothioacetamide in heterocyclic synthesis, see: Heterocycles, 24, 2023 (1986).