Ethyl palmitate, 97%
Ethyl palmitate, 97%
Ethyl palmitate, 97%
Thermo Scientific Chemicals

Ethyl palmitate, 97%

CAS: 628-97-7 | C18H36O2 | 284.48 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A15694.36
または、製品番号A15694-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS628-97-7
Appearance (Color)White or clear colorless
FormFused solid or liquid as melt
Refractive Index1.4375-1.4425 @ 20?C
Assay (GC)≥96.0%
Ethyl palmitate acts as a lubricant on the skin’s surfaces in order to get a soft and smooth appearance. It increases cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl palmitate acts as a lubricant on the skin’s surfaces in order to get a soft and smooth appearance. It increases cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol.

Solubility
Miscible with ethanol, dimethyl sulfoxide, chloroform and ethyl acetate. Immiscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Costa, M. C.; Boros, L. A. D.; Batista, M. L. S.; Coutinho, J. A. P.; Krähenbühl, M. A.; Meirelles, A. J. Phase diagrams of mixtures of ethyl palmitate with fatty acid ethyl esters. Fuel 2012 91 (1), 177-181.
  2. Kanda, L. R.; Voll, F. A. P.; Corazza, M. L. LLE for the systems ethyl palmitate (palmitic acid)(1)+ ethanol (2)+ glycerol (water)(3). Fluid Phase Equilib. 2013, 354, 147-155.