2-Chloroaniline, 98+%
2-Chloroaniline, 98+%
2-Chloroaniline, 98+%
Thermo Scientific Chemicals

2-Chloroaniline, 98+%

CAS: 95-51-2 | C6H6ClN | 127.571 g/mol
数量:
10,000 g
500 g
2500 g
製品番号(カタログ番号) A15758.36
または、製品番号A15758-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS95-51-2
IUPAC Name2-chloroaniline
Molecular FormulaC6H6ClN
InChI KeyAKCRQHGQIJBRMN-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1Cl
さらに表示
Appearance (Color)Clear colorless to yellow to orange to brown or pale orange-red
FormLiquid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Refractive Index1.5860-1.5900 @ 20°C
2-Chloroaniline used as petroleum solvents. It is used in biological studies to evaluate the mutagenicity and carcinogenicity of Salmonella typhimurium. Used as a Intermediates for dyestuff, pigment and specialty chemicals. 2-Chloroaniline is widely used in polymer, rubber, pharmaceutical and dye industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroaniline used as petroleum solvents. It is used in biological studies to evaluate the mutagenicity and carcinogenicity of Salmonella typhimurium. Used as a Intermediates for dyestuff, pigment and specialty chemicals. 2-Chloroaniline is widely used in polymer, rubber, pharmaceutical and dye industries.

Solubility
Miscible with alcohol, ether. Soluble in acid and most organic solvents. Very slightly soluble in water (0.5g/100mL at 20°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. V.A. Rana; A.D. Vyas; S.C. Mehrotra. Dielectric relaxation study of mixtures of 1-propanol with aniline, 2-chloroaniline and 3-chloroaniline at different temperatures using time domain reflectometry. Journal of Molecular Liquids. 2003, 102 (1-3), 379-391.
  2. Jen-Fon Jen; Chun-Tan Chang; Thomas C. Yang. On-line microdialysis-high-performance liquid chromatographic determination of aniline and 2-chloroaniline in polymer industrial wastewater. Journal of Chromatography A. 2001, 930 (1-2), 119-125.