3-Nitrobenzonitrile, 98%
3-Nitrobenzonitrile, 98%
3-Nitrobenzonitrile, 98%
Thermo Scientific Chemicals

3-Nitrobenzonitrile, 98%

CAS: 619-24-9 | C7H4N2O2 | 148.121 g/mol
数量:
10 g
25 g
250 g
製品番号(カタログ番号) A15821.09
または、製品番号A15821-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS619-24-9
IUPAC Name3-nitrobenzonitrile
Molecular FormulaC7H4N2O2
InChI KeyRUSAWEHOGCWOPG-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC(=C1)C#N
さらに表示
Melting Point (clear melt)113.0-119.0?C
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder or lumps or needles
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only