DL-Norvaline, 98%
DL-Norvaline, 98%
DL-Norvaline, 98%
Thermo Scientific Chemicals

DL-Norvaline, 98%

CAS: 760-78-1 | C5H11NO2 | 117.15 g/mol
製品番号(カタログ番号) A15900.22
または、製品番号A15900-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS760-78-1
IUPAC Name2-aminopentanoic acid
Molecular FormulaC5H11NO2
InChI KeySNDPXSYFESPGGJ-UHFFFAOYSA-N
SMILESCCCC(N)C(O)=O
さらに表示
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White
DL-Norvaline is a neurotransmitter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Norvaline is a neurotransmitter.

Solubility
Soluble in water.

Notes
Store in cool dry place.. Keep away from oxidizing agents.
RUO – Research Use Only
H. J. Svec.; D. D. Clyde. Vapor Pressures of Some α-Amino Acids. J. Chem. Eng. Data. 1965, 10 (2),151-152 .