N-Phthaloylglycine, 98+%
N-Phthaloylglycine, 98+%
N-Phthaloylglycine, 98+%
Thermo Scientific Chemicals

N-Phthaloylglycine, 98+%

CAS: 4702-13-0 | C10H7NO4 | 205.169 g/mol
数量:
50 g
250 g
1000 g
製品番号(カタログ番号) A15920.0B
または、製品番号A15920-0B
価格(JPY)
-
数量:
1000 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4702-13-0
IUPAC Name2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)acetic acid
Molecular FormulaC10H7NO4
InChI KeyWQINSVOOIJDOLJ-UHFFFAOYSA-N
SMILESOC(=O)CN1C(=O)C2=CC=CC=C2C1=O
さらに表示
Appearance (Color)White
Assay (Silylated GC)>98.0%
Assay (Aqueous acid-base Titration)>98.0%
Formpowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only