1-Bromo-4-tert-butylbenzene, 97%
1-Bromo-4-tert-butylbenzene, 97%
1-Bromo-4-tert-butylbenzene, 97%
Thermo Scientific Chemicals

1-Bromo-4-tert-butylbenzene, 97%

CAS: 3972-65-4 | C10H13Br | 213.118 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A15924.22
または、製品番号A15924-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3972-65-4
IUPAC Name1-bromo-4-tert-butylbenzene
Molecular FormulaC10H13Br
InChI KeyXHCAGOVGSDHHNP-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(Br)C=C1
さらに表示
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.5310-1.5350 @ 20?C
FormLiquid
1-Bromo-4-tert-butylbenzene was used in the synthesis of 4-tert-butyl-phenylboronic acid1, 1-deoxy analogs of CP-47,497 (n = 0 to 7) and 1-deoxy analogs of CP-55,940 (n = 0 to 7). It undergoes lithium-bromide exchange reactions with n-butyllithium and tert-butyllithium at 0°C in various solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-tert-butylbenzene was used in the synthesis of 4-tert-butyl-phenylboronic acid1, 1-deoxy analogs of CP-47,497 (n = 0 to 7) and 1-deoxy analogs of CP-55,940 (n = 0 to 7). It undergoes lithium-bromide exchange reactions with n-butyllithium and tert-butyllithium at 0°C in various solvents.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Michael Palucki.; Stephen L. Buchwald. Palladium-Catalyzed α-Arylation of Ketones.J. Am. Chem. Soc. 1997, 119 (45),11108-11109 .
  2. Jens Åhman.; John P. Wolfe.; Malisa V. Troutman.; Michael Palucki.; Stephen L. Buchwald. Asymmetric Arylation of Ketone Enolates.J. Am. Chem. Soc. 1998, 120 (8), 1918-1919 .