Tetrazolium Violet, 98+%
Tetrazolium Violet, 98+%
Tetrazolium Violet, 98+%
Thermo Scientific Chemicals

Tetrazolium Violet, 98+%

CAS: 1719-71-7 | C23H17ClN4 | 384.867 g/mol
製品番号(カタログ番号) A16008.14
または、製品番号A16008-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1719-71-7
IUPAC Name2-(naphthalen-1-yl)-3,5-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular FormulaC23H17ClN4
InChI KeyRONADMZTCCPLEF-UHFFFAOYSA-M
SMILES[Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=C2C=CC=CC2=CC=C1)C1=CC=CC=C1
さらに表示
FormPowder
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Appearance (Color)Cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only