(-)-Dibenzoyl-L-tartaric acid,98%
(-)-Dibenzoyl-L-tartaric acid,98%
(-)-Dibenzoyl-L-tartaric acid,98%
Thermo Scientific Chemicals

(-)-Dibenzoyl-L-tartaric acid,98%

CAS: 2743-38-6 | C18H14O8 | 358.30 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A16181.14
または、製品番号A16181-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2743-38-6
IUPAC Name(2R,3R)-2,3-bis(benzoyloxy)butanedioic acid
Molecular FormulaC18H14O8
InChI KeyYONLFQNRGZXBBF-ZIAGYGMSSA-N
SMILESOC(=O)[C@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O
さらに表示
Optical Rotation-120? ? 3? (c=5 in methanol)
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder or crystalline powder
(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
It is sensitive to moisture. Incompatible with oxidizing agents.
RUO – Research Use Only
Malachy McCann; Fergal Humphreys; Vickie McKee. Transition metal complexes of dibenzoyl-L-tartaric acid (db-L-tarH2) and L-tartaric acid (L-tarH2) ; X-ray crystal structure of {[Cu(L-tar) (phen)] · 6H2O}n (phen = 1,10-phenanthroline). Polyhedron. 1997, 16, (20),3655-3661