(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%
(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%
(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%
Thermo Scientific Chemicals

(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%

CAS: 1779-58-4 | C21H20BrO2P | 415.27 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A16301.14
または、製品番号A16301-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1779-58-4
IUPAC Name(2-methoxy-2-oxoethyl)triphenylphosphanium bromide
Molecular FormulaC21H20BrO2P
InChI KeyVCWBQLMDSMSVRL-UHFFFAOYSA-M
SMILES[Br-].COC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
FormPowder
Appearance (Color)White
Assay (Titration ex Bromide)≥98.0 to ≤102.0%
(Methoxycarbonylmethyl)triphenylphosphonium bromide is a reactant for the synthesis of GSK-3 inhibitors using the Wittig reaction, substituted furanones for antifungal and cytostatic activities, photochromic dithienylethene derivatives, gem-diiodoalkanes containing allylic alcohols and highly substituted cyclopentenes and brazilanes via Au-catalyzed deoxygenative Nazarov cyclization. As reactant for asymmetric hydrogenation of α,β-unsaturated ester-phosphonates, studies of thermal decomposition of phosphonium alkyl ester salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Methoxycarbonylmethyl)triphenylphosphonium bromide is a reactant for the synthesis of GSK-3 inhibitors using the Wittig reaction, substituted furanones for antifungal and cytostatic activities, photochromic dithienylethene derivatives, gem-diiodoalkanes containing allylic alcohols and highly substituted cyclopentenes and brazilanes via Au-catalyzed deoxygenative Nazarov cyclization. As reactant for asymmetric hydrogenation of α,β-unsaturated ester-phosphonates, studies of thermal decomposition of phosphonium alkyl ester salts.

Solubility
Soluble in methanol(almost transparency).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only

General References:

  1. Petr Šenel et. al. Antifungal 3,5-disubstituted furanones: From 5-acyloxymethyl to 5-alkylidene derivatives. Bioorganic & Medicinal Chemistry. 2010, 18 (5), 1988-2000.
  2. Oliver Tosic et. al.Photochromic dithienylethenes with extended pi-systems. Photochemical & Photobiological Sciences. 2010, 9,(2), 128-130.