3-Aminoquinoline, 98%
3-Aminoquinoline, 98%
3-Aminoquinoline, 98%
Thermo Scientific Chemicals

3-Aminoquinoline, 98%

CAS: 580-17-6 | C9H8N2 | 144.177 g/mol
製品番号(カタログ番号) A16592.03
または、製品番号A16592-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS580-17-6
IUPAC Namequinolin-3-amine
Molecular FormulaC9H8N2
InChI KeySVNCRRZKBNSMIV-UHFFFAOYSA-N
SMILESNC1=CN=C2C=CC=CC2=C1
さらに表示
Melting Point (clear melt)90.0-97.0°C
Assay (GC)≥97.5%
Appearance (Color)Pale cream or pale orange to pale brown to gray
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only