Bismarck Brown Y
Bismarck Brown Y
Bismarck Brown Y
Thermo Scientific Chemicals

Bismarck Brown Y

CAS: 10114-58-6 | C18H20Cl2N8 | 419.314 g/mol
製品番号(カタログ番号) A16674.22
または、製品番号A16674-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10114-58-6
IUPAC Namedihydrogen 4-(2-{3-[2-(2,4-diaminophenyl)diazen-1-yl]phenyl}diazen-1-yl)benzene-1,3-diamine dichloride
Molecular FormulaC18H20Cl2N8
InChI KeyMCZVRBLCRZWFJH-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NC1=CC=C(N=NC2=CC(=CC=C2)N=NC2=CC=C(N)C=C2N)C(N)=C1
さらに表示
Appearance (Color)Powder
Appearance (Color)Red-brown to dark brown or black
Assay (Dye)≥45%
CommentSpecification differs for U.S. and non-U.S. material where indicated
lambda-max (UV-Vis)454-464nm (in acidified 50% ethanol) (non-U.S. specification)
さらに表示
Plasma stain for mucins, cartilage and goblet cellsBismarck Brown Y is used for staining mucine, cartilage and goblet cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Plasma stain for mucins, cartilage and goblet cellsBismarck Brown Y is used for staining mucine, cartilage and goblet cells.

Solubility
Soluble in water. Slightly soluble in ethanol and cellosolve. Insoluble in acetone, carbon tetrachloride and benzene.

Notes
Incompatible with strong oxidizing agents and reducing agents.
RUO – Research Use Only

General References:

  1. Safarik, I.; Maderova, Z.; Horska, K.; Baldikova, E.; Pospiskova, K.; Safarikova, M. Spent Rooibos (Aspalathus linearis) Tea Biomass as an Adsorbent for Organic Dye Removal. Biorem. J. 2015, 19 (3), 183-187.
  2. Safarik, I.; Horska, K.; Pospiskova, K.; Filip, J.; Safarikova, M. Mechanochemical synthesis of magnetically responsive materials from non-magnetic precursors. Mater. Lett. 2014, 126, 202-206.