1,5-Dimethylnaphthalene, 99%
1,5-Dimethylnaphthalene, 99%
1,5-Dimethylnaphthalene, 99%
Thermo Scientific Chemicals

1,5-Dimethylnaphthalene, 99%

CAS: 571-61-9 | C12H12 | 156.23 g/mol
数量:
100 mg
500 mg
製品番号(カタログ番号) A16819.ME
または、製品番号A16819-ME
価格(JPY)
-
数量:
500 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS571-61-9
IUPAC Name1,5-dimethylnaphthalene
Molecular FormulaC12H12
InChI KeySDDBCEWUYXVGCQ-UHFFFAOYSA-N
SMILESCC1=CC=CC2=C(C)C=CC=C12
さらに表示
Appearance (Color)White
FormCrystals or powder or crystalline powder or flakes
Melting Point (clear melt)79.5-83.5?C
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only