N,N'-Dimethyl-1,3-propanediamine, 97%
N,N'-Dimethyl-1,3-propanediamine, 97%
N,N'-Dimethyl-1,3-propanediamine, 97%
Thermo Scientific Chemicals

N,N'-Dimethyl-1,3-propanediamine, 97%

CAS: 111-33-1 | C5H16N2 | 104.20 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A16901.14
または、製品番号A16901-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS111-33-1
IUPAC Namemethyl[3-(methylazaniumyl)propyl]azanium
Molecular FormulaC5H16N2
InChI KeyUQUPIHHYKUEXQD-UHFFFAOYSA-P
SMILESC[NH2+]CCC[NH2+]C
さらに表示
Refractive Index1.4360-1.4415 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
FormLiquid
Employed as a pharmaceutical intermediate. It is an important raw material used in organic synthesis, agrochemicals and dye stuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a pharmaceutical intermediate. It is an important raw material used in organic synthesis, agrochemicals and dye stuffs.

Solubility
Fully miscible in water, 1e+006 mg/L @ 25°C (est).

Notes
Air sensitive. Store away from oxidizing agents, carbon dioxide and air. Store under inert gas.
RUO – Research Use Only

General References:

  1. D. Danon.; L. Goldstein.; Y. Marikovsky.; E. Skutelsky. Use of cationized ferritin as a label of negative charges on cell surfaces.Journal of Ultrastructure Research,1972,38(5-6), 500-510.
  2. S K Basu.; J L Goldstein.; G W Anderson.; and M S Brown. Degradation of cationized low density lipoprotein and regulation of cholesterol metabolism in homozygous familial hypercholesterolemia fibroblasts.Proc. Nati. Acad. Sci.,1976,73(9), 3178-3182.