2-Fluorotoluene, 99+%
2-Fluorotoluene, 99+%
2-Fluorotoluene, 99+%
Thermo Scientific Chemicals

2-Fluorotoluene, 99+%

CAS: 95-52-3 | C7H7F | 110.131 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A16912.36
または、製品番号A16912-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS95-52-3
IUPAC Name1-fluoro-2-methylbenzene
Molecular FormulaC7H7F
InChI KeyMMZYCBHLNZVROM-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1F
さらに表示
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥99.0%
FormLiquid
Refractive Index1.4725-1.4745 @ 20?C
It is employed in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in organic synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

General References:

  1. Ted Schaefer.; Rudy Sebastian.; Richard P. Veregin.; Reino Laatikainen. Some unique properties of in 2-fluorotoluene derivatives. Conformational dependence.Can. J. Chem.,1983,61(1), 29-36.
  2. Sushil K. Maurya.; M. K. Gurjar.; K. M. Malshe.; P. T. Patil.; M. K. Dongare.; and Erhard Kemnitz. Solid acid catalysts for fluorotoluene nitration using nitric acid.Green Chem.,2003,5720-723.