Xylitol, 99%
Xylitol, 99%
Xylitol, 99%
Thermo Scientific Chemicals

Xylitol, 99%

CAS: 87-99-0 | C5H12O5 | 152.15 g/mol
製品番号(カタログ番号) A16944.36
または、製品番号A16944-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS87-99-0
IUPAC Namepentane-1,2,3,4,5-pentol
Molecular FormulaC5H12O5
InChI KeyHEBKCHPVOIAQTA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)CO
さらに表示
Appearance (Color)White
Identification (FTIR)Conforms
Assay (Silylated GC)≥98.5%
Melting Point91.5-97.5?C
FormCrystalline powder
It is employed as a polyol substrate for xylitol and sorbitol dehydrogenases and sorbitol dehydrogenase derived from xylose. It is applied as a sweetener and humectants and skin conditioning agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a polyol substrate for xylitol and sorbitol dehydrogenases and sorbitol dehydrogenase derived from xylose. It is applied as a sweetener and humectants and skin conditioning agent in cosmetics industry.

Solubility
Soluble in water (50 mg/ml), abs methanol (6 g/100ml), abs ethanol (1.2 g/100ml), alcohol (very slightly), and pyridine. Insoluble in ether.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Store under inert gas.
RUO – Research Use Only

General References:

  1. Amir Azarpazhooh.; Hardy Limeback.; Herenia P Lawrence.; Prakeshkumar S Shah. Xylitol for preventing acute otitis media in children up to 12 years of age.Cochrane Database Syst Rev.2011,11.
  2. Trahan L. Xylitol: a review of its action on mutans streptococci and dental plaque--its clinical significance.International Dental Journal,1995,45.(1), 77-92.