o-Dianisidine dihydrochloride, 98%
o-Dianisidine dihydrochloride, 98%
o-Dianisidine dihydrochloride, 98%
Thermo Scientific Chemicals

o-Dianisidine dihydrochloride, 98%

CAS: 20325-40-0 | C14H18Cl2N2O2 | 317.21 g/mol
製品番号(カタログ番号) A17175.03
または、製品番号A17175-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS20325-40-0
IUPAC Namedihydrogen 3,3'-dimethoxy-[1,1'-biphenyl]-4,4'-diamine dichloride
Molecular FormulaC14H18Cl2N2O2
InChI KeyUXTIAFYTYOEQHV-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N)C1=CC=C(N)C(OC)=C1
さらに表示
Appearance (Color)White to cream to grey to purple-grey
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5% (Dry wt. basis)
Loss on Drying≤5%
It is suitable for use in ELISA procedures. In neurohistochemistry o-Dianisidine dihydrochloride has demonstrated perikaryal labeling as well as retrograde and anterograde transport to the primary olfactory cortex. It has also been used for a paper based-micro fluidic device to semi-quantitatively measure lactate, uric acid, and glucose. 3,3'-Dimethoxybenzidine is used principally as an intermediate in the production of commercial bisazobiphenyl dyes for coloring textiles, paper, plastic, rubber, and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is suitable for use in ELISA procedures. In neurohistochemistry o-Dianisidine dihydrochloride has demonstrated perikaryal labeling as well as retrograde and anterograde transport to the primary olfactory cortex. It has also been used for a paper based-micro fluidic device to semi-quantitatively measure lactate, uric acid, and glucose. 3,3′-Dimethoxybenzidine is used principally as an intermediate in the production of commercial bisazobiphenyl dyes for coloring textiles, paper, plastic, rubber, and leather.

Solubility
Soluble in water (50 mg/ml). It dissolves in ethyl alcohol ,acid and in organic solvent.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only

General References:

  1. Yanfen Peng.; Zhiqiang Gao.Amplified detection of microRNA based on ruthenium oxide nanoparticle-initiated deposition of an insulating film.Anal Chem.2011,83(3), 820-827.
  2. Wei Shen.; Huimin Deng.; Yuqian Ren, Zhiqiang Gao. A label-free microRNA biosensor based on DNAzyme-catalyzed and microRNA-guided formation of a thin insulating polymer film.Biosens Bioelectron.2013,44,171-176 .