4-n-Decylbenzoyl chloride, 98%
4-n-Decylbenzoyl chloride, 98%
4-n-Decylbenzoyl chloride, 98%
Thermo Scientific Chemicals

4-n-Decylbenzoyl chloride, 98%

CAS: 54256-43-8 | C17H25ClO | 280.836 g/mol
製品番号(カタログ番号) A17234.06
または、製品番号A17234-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS54256-43-8
IUPAC Name4-decylbenzoyl chloride
Molecular FormulaC17H25ClO
InChI KeyHEWLDHSFOQXCSI-UHFFFAOYSA-N
SMILESCCCCCCCCCCC1=CC=C(C=C1)C(Cl)=O
さらに表示
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5115-1.5155 @ 20?C
Assay (GC)≥97.5%
FormLiquid
Employed as a pharmaceutical and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a pharmaceutical and chemical intermediate.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas.
RUO – Research Use Only
Yonghong Gan.; Pingzhen Wang.; and Thomas A. Spencer. Synthesis of Benzophenone-Containing Fatty Acids.J. Org. Chem.,2006,71,(25), 9487-9490.