Metanil Yellow
Metanil Yellow
Metanil Yellow
Thermo Scientific Chemicals

Metanil Yellow

CAS: 587-98-4 | C18H14N3NaO3S | 375.378 g/mol
製品番号(カタログ番号) A17527.30
または、製品番号A17527-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS587-98-4
IUPAC Namesodium 3-{2-[4-(phenylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
Molecular FormulaC18H14N3NaO3S
InChI KeyNYGZLYXAPMMJTE-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C1=CC=CC(=C1)N=NC1=CC=C(NC2=CC=CC=C2)C=C1
さらに表示
Specific Absorbance (E 1%/1cm)≥300 at lambda max (non-U.S. sourced material)
CommentMaterial sourced in the U.S. and in other countries
lambda-max (UV-Vis)431-438nm (water, non-U.S. sourced material); 410-420 nm (methanol, U.S. sourced material)
Appearance (Color)Yellow to orange
FormPowder
さらに表示
Metanil Yellow is used as indicator. Metanil yellow is useful in spectrophotometric methods to determine the presence of trace amounts of nitrates and nitrites in solution. The method utilizes the fact that potassium bromate can catalyze the discoloring of Metanil yellow which can then be analyzed by spectrophotometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metanil Yellow is used as indicator. Metanil yellow is useful in spectrophotometric methods to determine the presence of trace amounts of nitrates and nitrites in solution. The method utilizes the fact that potassium bromate can catalyze the discoloring of Metanil yellow which can then be analyzed by spectrophotometry.

Solubility
Soluble in water (25g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. T.N. Nagaraja.; T. Desiraju. Effects of chronic consumption of metanil yellow by developing and adult rats on brain regional levels of noradrenaline, dopamine and serotonin, on acetylcholine esterase activity and on operant conditioning. Food and Chemical Toxicology. 1993, 31, (1), 41-44.
  2. Ming-Shen Chiou.; Guo-Syong Chuang. Competitive adsorption of dye metanil yellow and RB15 in acid solutions on chemically cross-linked chitosan beads. Chemosphere. 2006, 62, (5), 731-740.