DL-Malic acid, 98%
DL-Malic acid, 98%
DL-Malic acid, 98%
Thermo Scientific Chemicals

DL-Malic acid, 98%

CAS: 6915-15-7 | C4H6O5 | 134.087 g/mol
製品番号(カタログ番号) A17874.0E
または、製品番号A17874-0E
価格(JPY)
-
数量:
2500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6915-15-7
IUPAC Name2-hydroxybutanedioic acid
Molecular FormulaC4H6O5
InChI KeyBJEPYKJPYRNKOW-UHFFFAOYNA-N
SMILESOC(CC(O)=O)C(O)=O
さらに表示
Optical Rotation-0.5? to +0.5? (c= 8.5 in water)
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
FormCrystalline powder or crystals
さらに表示
DL-Malic acid is used as a food additive. It is a component of some artificial vinegar flavors. Further, it is used in the preparation of chiral compounds, including κ-opioid rece. It is utilized for chiral resolution by ligand-exchange capillary electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Malic acid is used as a food additive. It is a component of some artificial vinegar flavors. Further, it is used in the preparation of chiral compounds, including κ-opioid rece. It is utilized for chiral resolution by ligand-exchange capillary electrophoresis.

Solubility
Soluble in acetone, ether, water, methanol and ethanol.

Notes
Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
RUO – Research Use Only

General References:

  1. Zhang, Q.; Yang, Y.; Cheng, L.; Cao, C.; Ding, Z.; Wang, C.; Yang, W.; Hu, Y.; Li, Y. Thermodynamic models for determination of the solubility of dl-malic acid in methanol plus (acetonitrile, N,N-dimethylformamide, isopropyl alcohol) binary solvent mixtures. J. Chem. Thermodyn. 2015, 85, 148-154.
  2. Espinosa, K. C.; Cabrera, R. I. G.; López, A. B.; Roldán, M. A. T.; Cruz, N. A. V. Positive effect of reduced aeration rate on growth and stereospecificity of dl-malic acid consumption by Azospirillum brasilense: Improving the shelf life of a liquid inoculant formulation. J. Biotechnol. 2015, 195, 74-81.