Isonicotinamide, 99%
Isonicotinamide, 99%
Isonicotinamide, 99%
Thermo Scientific Chemicals

Isonicotinamide, 99%

CAS: 1453-82-3 | C6H6N2O | 122.127 g/mol
数量:
50 g
100 g
250 g
500 g
製品番号(カタログ番号) A18190.30
または、製品番号A18190-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1453-82-3
IUPAC Namepyridine-4-carboxamide
Molecular FormulaC6H6N2O
InChI KeyVFQXVTODMYMSMJ-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=NC=C1
さらに表示
Identification (FTIR)Conforms
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)153.0-160.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only